APO455831436 Apollo Scientific Ltd

1-[4-(Cyclopropylamino)piperidin-1-yl]ethan-1-one

  • MDL number MFCD01941331
  • Cas Number : 387358-46-5
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-acetyl-N-cyclopropyl-4-piperidinamine
Inchi Code 1S/C10H18N2O/c1-8(13)12-6-4-10(5-7-12)11-9-2-3-9/h9-11H,2-7H2,1H3
Inchi Key KWLJTPCIXGQHSB-UHFFFAOYSA-N

Description

Synonym 1-Acetyl-4-(cyclopropylamino)piperidine, [(1-Acetylpiperidin-4-yl)amino]cyclopropane
Purity 95%
Flash Point 120/0.075mm
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-cyclopropylaminopiperidin-1-ylethan-1-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-cyclopropylaminopiperidin-1-ylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart