APO455831443 Apollo Scientific Ltd

2-Methyl-1,8-naphthyridine-3-carboxylic acid

  • MDL number MFCD00202882
  • Cas Number : 387350-60-9
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-195
Synonym 3-Carboxy-2-methyl-1,8-naphthyridine, 1,8-Diaza-2-methylnaphthalene-3-carboxylic acid
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-18-naphthyridine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-18-naphthyridine-3-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart