APO455831482 Apollo Scientific Ltd

(Pyridin-3-yl)acetic acid

  • MDL number MFCD00066302
  • Cas Number : 501-81-5
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Synonym 3-(Carboxymethyl)pyridine
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/pyridin-3-ylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyridin-3-ylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart