APO455831490 Apollo Scientific Ltd

Ethyl 5-bromonicotinate

  • MDL number MFCD00040366
  • Cas Number : 20986-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Synonym Ethyl 5-bromopyridine-3-carboxylate
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 86-92/0.5mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13364

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13364
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart