APO455831497 Apollo Scientific Ltd

Methyl 4-(bromomethyl)phenylacetate

  • MDL number MFCD07783030
  • Cas Number : 7398-42-7
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 37-39
Iupac Name methyl [4-(bromomethyl)phenyl]acetate
Inchi Code 1S/C10H11BrO2/c1-13-10(12)6-8-2-4-9(7-11)5-3-8/h2-5H,6-7H2,1H3
Inchi Key APXOMRFLJBRHNX-UHFFFAOYSA-N

Description

Synonym 4-(2-Methoxy-2-oxoethyl)benzyl bromide, 4-(Bromomethyl)phenylacetic acid methyl ester
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 37-39
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromomethylphenylacetate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromomethylphenylacetate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart