APO455831507 Apollo Scientific Ltd

1-(tert-Butyl)piperazine

  • MDL number MFCD00049359
  • Cas Number : 38216-72-7
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 32-40
Iupac Name 1-tert-butylpiperazine
Inchi Code 1S/C8H18N2/c1-8(2,3)10-6-4-9-5-7-10/h9H,4-7H2,1-3H3
Inchi Key PVMNSAIKFPWDQG-UHFFFAOYSA-N

Description

Purity 98%
Flash Point 66-75/12mm
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 32-40
Product Link https://store.apolloscientific.co.uk/product/1-tert-butylpiperazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-tert-butylpiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart