APO455831511 Apollo Scientific Ltd

2-Chloro-4-methylbenzoic acid

  • MDL number MFCD03094667
  • Cas Number : 7697-25-8
  • Molecular Weight : 170.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Carboxy-3-chlorotoluene, 2-Chloro-p-toluic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart