APO455831512 Apollo Scientific Ltd

4-(Methylsulphonyl)aniline

  • MDL number MFCD00025353
  • Cas Number : 5470-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-138
Synonym 4-Aminophenyl methyl sulphone, 4-Mesylaniline
Density 1.44
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart