APO455831517 Apollo Scientific Ltd

3-Chloro-5-hydroxybenzonitrile

  • MDL number MFCD11226540
  • Cas Number : 473923-97-6
  • Molecular Weight : 153.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175(dec.)
Synonym 3-Chloro-5-cyanophenol
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-hydroxybenzonitrile
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart