APO455831524 Apollo Scientific Ltd

Methyl 5-hydroxypyridine-2-carboxylate

  • MDL number MFCD05256426
  • Cas Number : 30766-12-2
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 191-197
Iupac Name methyl 5-hydroxy-2-pyridinecarboxylate
Inchi Code 1S/C7H7NO3/c1-11-7(10)6-3-2-5(9)4-8-6/h2-4,9H,1H3
Inchi Key YYAYXDDHGPXWTA-UHFFFAOYSA-N

Description

Synonym Methyl 5-hydroxypicolinate, 5-Hydroxy-2-(methoxycarbonyl)pyridine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 191-197
Product Link https://store.apolloscientific.co.uk/product/methyl-5-hydroxypyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-hydroxypyridine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart