APO455831530 Apollo Scientific Ltd

5-Bromo-3-hydroxy-2-methoxypyridine

  • MDL number MFCD18256269
  • Cas Number : 1211589-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-methoxypyridin-3-ol, 5-Bromo-3-hydroxypyridin-2-yl methyl ether
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-hydroxy-2-methoxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-hydroxy-2-methoxypyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart