APO455831534 Apollo Scientific Ltd

3-Bromo-5-nitrobenzyl alcohol

  • MDL number MFCD09056873
  • Cas Number : 139194-79-9
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 88-90
Iupac Name (3-bromo-5-nitrophenyl)methanol
Inchi Code 1S/C7H6BrNO3/c8-6-1-5(4-10)2-7(3-6)9(11)12/h1-3,10H,4H2
Inchi Key WIXGOPRKQQGTHO-UHFFFAOYSA-N

Description

Synonym (3-Bromo-5-nitrophenyl)methanol, 3-Bromo-5-(hydroxymethyl)nitrobenzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 88-90
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-nitrobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart