APO455831535 Apollo Scientific Ltd

2,5,6-Triaminopyrimidin-4(1H)-one sulphate

  • MDL number MFCD27664806
  • Cas Number : 39267-74-8
  • Molecular Weight : 239.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sulfuric acid compound with 2,5,6-triamino-4-pyrimidinol (1:1)
Inchi Code 1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4)
Inchi Key RSKNEEODWFLVFF-UHFFFAOYSA-N

Description

Synonym 2,5,6-Triaminopyrimidin-4(3H)-one sulphate, 4-Hydroxy-2,5,6-triaminopyrimidine sulphate, 6-Hydroxy-2,4,5-triaminopyrimidine sulphate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/256-triaminopyrimidin-41h-one-sulphate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/256-triaminopyrimidin-41h-one-sulphate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart