APO455831539 Apollo Scientific Ltd

2-Iodo-6-methylbenzyl alcohol

  • MDL number MFCD18397812
  • Cas Number : 1261794-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60199

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=60199
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart