APO455831555 Apollo Scientific Ltd

2-Amino-4-bromobenzonitrile

  • MDL number MFCD08752638
  • Cas Number : 304858-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-163
Synonym 5-Bromo-2-cyanoaniline, 4-Bromoanthranilonitrile
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50049

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50049
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart