APO455831560 Apollo Scientific Ltd

8-Aminoimidazo[1,2-a]pyridine

  • MDL number MFCD06739233
  • Cas Number : 73221-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Imidazo[1,2-a]pyridin-8-amine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-aminoimidazo12-apyridine

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/8-aminoimidazo12-apyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart