APO455831561 Apollo Scientific Ltd

3'-Bromo-2'-methoxyacetophenone

  • MDL number MFCD22573042
  • Cas Number : 267651-23-0
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromo-2-methoxyphenyl)ethanone
Inchi Code 1S/C9H9BrO2/c1-6(11)7-4-3-5-8(10)9(7)12-2/h3-5H,1-2H3
Inchi Key IBHCVANJIBGASI-UHFFFAOYSA-N

Description

Synonym 1-(3-Bromo-2-methoxyphenyl)ethan-1-one, 2-Acetyl-6-bromoanisole
Purity 97%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-methoxyacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-methoxyacetophenone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart