APO455831565 Apollo Scientific Ltd

Methyl 3,5-dichloro-2-hydroxybenzoate

  • MDL number MFCD00464939
  • Cas Number : 7606-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Synonym Methyl 3,5-dichlorosalicylate, 2,4-Dichloro-6-(methoxycarbonyl)phenol
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51485

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51485
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart