APO455831590 Apollo Scientific Ltd

3-Bromo-4-nitroaniline

  • MDL number MFCD07783047
  • Cas Number : 40787-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-177
Synonym 4-Amino-2-bromonitrobenzene
Purity 99
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-nitroaniline

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-nitroaniline
pictograms GHS07
hstatements H312+H332;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart