APO455831613 Apollo Scientific Ltd

5-Chloro-2-ethynylpyridine

  • MDL number MFCD13189715
  • Cas Number : 1196153-33-9
  • Molecular Weight : 137.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Chloropyridin-2-yl)acetylene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-ethynylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-ethynylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart