APO455831615 Apollo Scientific Ltd

3-Bromo-5-ethynylpyridine

  • MDL number MFCD10698137
  • Cas Number : 866683-52-5
  • Molecular Weight : 182.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-ethynylpyridine
Inchi Code 1S/C7H4BrN/c1-2-6-3-7(8)5-9-4-6/h1,3-5H
Inchi Key KMTYILRNJPSUDX-UHFFFAOYSA-N

Description

Synonym (5-Bromopyridin-3-yl)acetylene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-ethynylpyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-ethynylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart