APO455831623 Apollo Scientific Ltd

6-Ethynylimidazo[1,2-a]pyrazine

  • MDL number MFCD21647858
  • Cas Number : 1372096-41-7
  • Molecular Weight : 143.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-ethynylimidazo[1,2-a]pyrazine
Inchi Code 1S/C8H5N3/c1-2-7-6-11-4-3-9-8(11)5-10-7/h1,3-6H
Inchi Key ALNLKKWQOAJCJY-UHFFFAOYSA-N

Description

Synonym (Imidazo[1,2-a]pyrazin-6-yl)acetylene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-ethynylimidazo12-apyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-ethynylimidazo12-apyrazine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart