APO455831624 Apollo Scientific Ltd

2-Chloro-3-ethynylpyrazine

  • MDL number MFCD19689588
  • Cas Number : 1374115-59-9
  • Molecular Weight : 138.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-3-ethynylpyrazine
Inchi Code 1S/C6H3ClN2/c1-2-5-6(7)9-4-3-8-5/h1,3-4H
Inchi Key JUAUUZGANIEVSA-UHFFFAOYSA-N

Description

Synonym 2-Chloro-3-ethynyl-1,4-diazine, (3-Chloropyrazin-2-yl)acetylene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-ethynylpyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-ethynylpyrazine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart