APO455831629 Apollo Scientific Ltd

2-Ethynylpyrazine

  • MDL number MFCD13189804
  • Cas Number : 153800-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Ethynyl-1,4-diazine, (Pyrazin-2-yl)acetylene
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-ethynylpyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-ethynylpyrazine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart