APO455831633 Apollo Scientific Ltd

5-Ethynylimidazo[1,2-a]pyridine

  • MDL number MFCD22199325
  • Cas Number : 1374115-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Imidazo[1,2-a]pyridin-5-yl)acetylene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50299

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50299
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart