APO455831676 Apollo Scientific Ltd

5-Chloro-3-methylbenzo[b]thiophene-2-sulphonyl chloride

  • MDL number MFCD00173797
  • Cas Number : 166964-33-6
  • Molecular Weight : 281.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 153
Iupac Name 5-chloro-3-methyl-1-benzothiophene-2-sulfonyl chloride
Inchi Code 1S/C9H6Cl2O2S2/c1-5-7-4-6(10)2-3-8(7)14-9(5)15(11,12)13/h2-4H,1H3
Inchi Key JLTUANWNGKWRQO-UHFFFAOYSA-N

Description

Synonym 5-Chloro-3-methyl-1-benzothiophene-2-sulphonyl chloride, 5-Chloro-2-(chlorosulphonyl)-3-methyl-1-benzothiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 153
Product Link https://store.apolloscientific.co.uk/product/5-chloro-3-methylbenzobthiophene-2-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-3-methylbenzobthiophene-2-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart