APO455831677 Apollo Scientific Ltd

5-Bromo-3-methylbenzo[b]thiophene-2-sulphonyl chloride

  • MDL number MFCD03094671
  • Cas Number : 338797-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-(chlorosulphonyl)-3-methylbenzo[b]thiophene, 5-Bromo-3-methyl-1-benzothiophene-2-sulphonyl chloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13335

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13335
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart