APO455831695 Apollo Scientific Ltd

[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid dihydrate

  • MDL number MFCD03410262
  • Cas Number : 1049785-94-5
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(tert-butoxycarbonyl)-1-piperazinyl]acetic acid dihydrate
Inchi Code 1S/C11H20N2O4.2H2O/c1-11(2,3)17-10(16)13-6-4-12(5-7-13)8-9(14)15;;/h4-8H2,1-3H3,(H,14,15);2*1H2
Inchi Key DGAIWMBOPSWHBS-UHFFFAOYSA-N

Description

Synonym tert-Butyl 4-(carboxymethyl)piperazine-1-carboxylate dihydrate, 2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]ethanoic acid dihydrate
Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylpiperazin-1-ylacetic-acid-dihydrate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tert-butoxycarbonylpiperazin-1-ylacetic-acid-dihydrate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart