APO455831707 Apollo Scientific Ltd

Methyl 4-chloro-3-methylbenzoate

  • MDL number MFCD00151838
  • Cas Number : 91367-05-4
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-chloro-3-methylbenzoate
density 1.21
Inchi Code 1S/C9H9ClO2/c1-6-5-7(9(11)12-2)3-4-8(6)10/h3-5H,1-2H3
Inchi Key QOTGNXMPQNGYDM-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-(methoxycarbonyl)toluene, Methyl 4-chloro-m-toluate
Density 1.21
Flash Point 78/0.06mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart