APO455831712 Apollo Scientific Ltd

2,6-Dimethylisonicotinic acid

  • MDL number MFCD03731176
  • Cas Number : 54221-93-1
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 276-291
Iupac Name 2,6-dimethylisonicotinic acid
Inchi Code 1S/C8H9NO2/c1-5-3-7(8(10)11)4-6(2)9-5/h3-4H,1-2H3,(H,10,11)
Inchi Key JRJLLMLYUFAZOM-UHFFFAOYSA-N

Description

Synonym 2,6-Dimethylpyridine-4-carboxylic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 276-291
Product Link https://store.apolloscientific.co.uk/product/26-dimethylisonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dimethylisonicotinic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart