APO455831715 Apollo Scientific Ltd

2,5-Dichloro-4-hydroxypyridine

  • MDL number MFCD01861982
  • Cas Number : 847664-65-7
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 184
Iupac Name 2,5-dichloro-4-pyridinol
Inchi Code 1S/C5H3Cl2NO/c6-3-2-8-5(7)1-4(3)9/h1-2H,(H,8,9)
Inchi Key RJKYZTVQNJPAGX-UHFFFAOYSA-N

Description

Synonym 2,5-Dichloropyridin-4-ol
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 184
Product Link https://store.apolloscientific.co.uk/product/25-dichloro-4-hydroxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-dichloro-4-hydroxypyridine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart