APO455831718 Apollo Scientific Ltd

3-Chlorophthalic anhydride

  • MDL number MFCD00023107
  • Cas Number : 117-21-5
  • Molecular Weight : 182.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 123-126
Iupac Name 4-chloro-2-benzofuran-1,3-dione
Inchi Code 1S/C8H3ClO3/c9-5-3-1-2-4-6(5)8(11)12-7(4)10/h1-3H
Inchi Key UERPUZBSSSAZJE-UHFFFAOYSA-N

Description

Synonym 4-Chloro-2-benzofuran-1,3-dione, 4-Chloroisobenzofuran-1,3-dione
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 123-126
Product Link https://store.apolloscientific.co.uk/product/3-chlorophthalic-anhydride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chlorophthalic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart