APO455831720 Apollo Scientific Ltd

3'-Bromo-2'-hydroxyacetophenone

  • MDL number MFCD08236725
  • Cas Number : 1836-05-1
  • Molecular Weight : 215.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-44
Synonym 1-(3-Bromo-2-hydroxyphenyl)ethan-1-one, 2-Acetyl-6-bromophenol
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 140-145/6mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-hydroxyacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-hydroxyacetophenone
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart