APO455831722 Apollo Scientific Ltd

4-(3-Nitrophenyl)morpholine

  • MDL number MFCD03195994
  • Cas Number : 116922-22-6
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 106-110
Iupac Name 4-(3-nitrophenyl)morpholine
Inchi Code 1S/C10H12N2O3/c13-12(14)10-3-1-2-9(8-10)11-4-6-15-7-5-11/h1-3,8H,4-7H2
Inchi Key DKECFAGKGYRYAT-UHFFFAOYSA-N

Description

Synonym 1-(Morpholin-4-yl)-3-nitrobenzene, 3-(Morpholin-4-yl)nitrobenzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 106-110
Product Link https://store.apolloscientific.co.uk/product/4-3-nitrophenylmorpholine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-3-nitrophenylmorpholine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart