APO455831726 Apollo Scientific Ltd

1-Methyl-1H-pyrazole-4-sulphonyl chloride

  • MDL number MFCD04968667
  • Cas Number : 288148-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-48
Synonym 4-(Chlorosulphonyl)-1-methyl-1H-pyrazole
Storage temp. Store at -20
Physical Form Solid-Crystals
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-4-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-methyl-1h-pyrazole-4-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart