APO455831728 Apollo Scientific Ltd

4-Bromo-2-methylbenzyl alcohol

  • MDL number MFCD11847398
  • Cas Number : 17100-58-2
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 70-75
Iupac Name (4-bromo-2-methylphenyl)methanol
Inchi Code 1S/C8H9BrO/c1-6-4-8(9)3-2-7(6)5-10/h2-4,10H,5H2,1H3
Inchi Key IFKWLKCPUIQXPU-UHFFFAOYSA-N

Description

Synonym (4-Bromo-2-methylphenyl)methanol, 5-Bromo-2-(hydroxymethyl)toluene
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70-75
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart