APO455831777 Apollo Scientific Ltd

5-Hydroxypyridine-2-carboxylic acid

  • MDL number MFCD00661309
  • Cas Number : 15069-92-8
  • Molecular Weight : 139.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 260-264
Iupac Name 5-hydroxy-2-pyridinecarboxylic acid
Inchi Code 1S/C6H5NO3/c8-4-1-2-5(6(9)10)7-3-4/h1-3,8H,(H,9,10)
Inchi Key CGRRUFNHHQCLDZ-UHFFFAOYSA-N

Description

Synonym 5-Hydroxypicolinic acid, 2-Carboxy-5-hydroxypyridine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 260-264
Product Link https://store.apolloscientific.co.uk/product/5-hydroxypyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxypyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart