APO455831778 Apollo Scientific Ltd

5-Methyl-1H-indole-2-boronic acid, N-BOC protected

  • MDL number MFCD04114580
  • Cas Number : 475102-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-116
Synonym 2-Borono-1-(tert-butoxycarbonyl)-5-methyl-1H-indole, tert-Butyl 2-borono-5-methyl-1H-indole-1-carboxylate
Purity 97
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Ship on Ice
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30918

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30918
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart