APO455831779 Apollo Scientific Ltd

5-Cyano-1H-indole-2-boronic acid, N-BOC protected

  • MDL number MFCD04114578
  • Cas Number : 475102-15-9
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 260-270
Iupac Name 1-(tert-butoxycarbonyl)-5-cyano-1H-indol-2-ylboronic acid
Inchi Code 1S/C14H15BN2O4/c1-14(2,3)21-13(18)17-11-5-4-9(8-16)6-10(11)7-12(17)15(19)20/h4-7,19-20H,1-3H3
Inchi Key AJWYBPNMPKTRDV-UHFFFAOYSA-N

Description

Synonym 1-(tert-Butoxycarbonyl)-5-cyano-1H-indole-2-boronic acid
Purity 98%
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 260-270
Product Link https://store.apolloscientific.co.uk/product/5-cyano-1h-indole-2-boronic-acid-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P301+P312;P337+P313
MSDS https://store.apolloscientific.co.uk/product/5-cyano-1h-indole-2-boronic-acid-n-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart