APO455831786 Apollo Scientific Ltd

4-Bromopyridine-2-carboxylic acid

  • MDL number MFCD03426665
  • Cas Number : 30766-03-1
  • Molecular Weight : 202.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-174
Synonym 4-Bromopicolinic acid, 4-Bromo-2-carboxypyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromopyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromopyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart