APO455831791 Apollo Scientific Ltd

2-Amino-3-bromo-5-nitropyridine

  • MDL number MFCD00955628
  • Cas Number : 15862-31-4
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 218-222
Iupac Name 3-bromo-5-nitro-2-pyridinylamine
Inchi Code 1S/C5H4BrN3O2/c6-4-1-3(9(10)11)2-8-5(4)7/h1-2H,(H2,7,8)
Inchi Key OFXNHXMPRZDIDM-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-nitropyridin-2-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 218-222
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-bromo-5-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart