APO455831798 Apollo Scientific Ltd

6-Acetamidonicotinic acid

  • MDL number MFCD00129118
  • Cas Number : 21550-48-1
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 276-278
Iupac Name 6-(acetylamino)nicotinic acid
Inchi Code 1S/C8H8N2O3/c1-5(11)10-7-3-2-6(4-9-7)8(12)13/h2-4H,1H3,(H,12,13)(H,9,10,11)
Inchi Key RXSLHYTZMIUANX-UHFFFAOYSA-N

Description

Synonym 6-(Acetylamino)nicotinic acid, 6-(Acetylamino)pyridine-3-carboxylic acid
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 276-278
Product Link https://store.apolloscientific.co.uk/product/6-acetamidonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-acetamidonicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart