APO455831805 Apollo Scientific Ltd

2-Amino-1,3-thiazole-4-carboxylic acid, 2-BOC protected

  • MDL number MFCD02181057
  • Cas Number : 83673-98-7
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 250-252
Iupac Name 2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-4-carboxylic acid
Inchi Code 1S/C9H12N2O4S/c1-9(2,3)15-8(14)11-7-10-5(4-16-7)6(12)13/h4H,1-3H3,(H,12,13)(H,10,11,14)
Inchi Key PIWSRJPUYPNQJE-UHFFFAOYSA-N

Description

Synonym 2-[(tert-Butoxycarbonyl)amino]-1,3-thiazole-4-carboxylic acid, 2-[(tert-Butoxycarbonyl)amino]-4-carboxy-1,3-thiazole
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 250-252
Product Link https://store.apolloscientific.co.uk/product/2-amino-13-thiazole-4-carboxylic-acid-2-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-13-thiazole-4-carboxylic-acid-2-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart