APO455831812 Apollo Scientific Ltd

Ethyl 2-amino-1,3-oxazole-5-carboxylate

  • MDL number MFCD06657433
  • Cas Number : 113853-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-153
Synonym 2-Amino-5-(ethoxycarbonyl)-1,3-oxazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-oxazole-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-oxazole-5-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart