APO455831815 Apollo Scientific Ltd

Methyl 2-amino-5-bromo-1,3-thiazole-4-carboxylate

  • MDL number MFCD00137990
  • Cas Number : 850429-60-6
  • Molecular Weight : 237.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 157-159
Iupac Name methyl 2-amino-5-bromo-1,3-thiazole-4-carboxylate
Inchi Code 1S/C5H5BrN2O2S/c1-10-4(9)2-3(6)11-5(7)8-2/h1H3,(H2,7,8)
Inchi Key KVUHCAXYWHQFLW-UHFFFAOYSA-N

Description

Synonym 2-Amino-5-bromo-4-(methoxycarbonyl)-1,3-thiazole
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 157-159
Product Link https://store.apolloscientific.co.uk/product/methyl-2-amino-5-bromo-13-thiazole-4-carboxylate

Safety Information

signalword Warning
pcodes P280;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-2-amino-5-bromo-13-thiazole-4-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart