APO455831817 Apollo Scientific Ltd

Ethyl 2-amino-1,3-thiazole-5-carboxylate, 2-BOC protected

  • MDL number MFCD06409216
  • Cas Number : 302964-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-200
Synonym 2-[(tert-Butoxycarbonyl)amino]-5-(ethoxycarbonyl)-1,3-thiazole, Ethyl 2-[(tert-butoxycarbonyl)amino]-1,3-thiazole-5-carboxylate
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-thiazole-5-carboxylate-2-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-13-thiazole-5-carboxylate-2-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart