APO455831819 Apollo Scientific Ltd

5-(Chloroacetyl)-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD04115734
  • Cas Number : 65435-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228-232
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloroacetyl-13-dihydro-2h-indol-2-one

Safety Information

signalword Warning
pcodes P280;P260;P261;P271
MSDS https://store.apolloscientific.co.uk/product/5-chloroacetyl-13-dihydro-2h-indol-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart