APO455831820 Apollo Scientific Ltd

3-Bromo-5-nitrotoluene

  • MDL number MFCD06659815
  • Cas Number : 52488-28-5
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 81-84
Iupac Name 1-bromo-3-methyl-5-nitrobenzene
Inchi Code 1S/C7H6BrNO2/c1-5-2-6(8)4-7(3-5)9(10)11/h2-4H,1H3
Inchi Key MWFDNXJPZUOTJB-UHFFFAOYSA-N

Description

Synonym 1-Bromo-3-methyl-5-nitrobenzene, 3-Bromo-5-methylnitrobenzene
Purity 96%
Flash Point 269-270
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 81-84
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-nitrotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-nitrotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart