APO455831821 Apollo Scientific Ltd

trans-4-Aminocyclohexanol

  • MDL number MFCD00067698
  • Cas Number : 27489-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-113
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 124/14mm
Product Link https://store.apolloscientific.co.uk/product/trans-4-aminocyclohexanol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-4-aminocyclohexanol
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart