APO455831836 Apollo Scientific Ltd

2,6-Bis(dimethylamino)pyrimidine-4-carboxylic acid monohydrate

  • MDL number MFCD07366474
  • Cas Number : 1052402-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-168
Synonym 2,4-Bis(dimethylamino)pyrimidine-6-carboxylic acid monohydrate, 2,6-Bis(dimethylamino)-1,3-diazine-4-carboxylic acid monohydrate
Purity 95
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13114

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13114
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart